The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000108.4 | NP_000099.2 | 10 - 11 | 1191 | 117 |
NM_001289750.1 | NP_001276679.1 | 8 - 9 | 1042 | 117 | |
NM_001289751.1 | NP_001276680.1 | 9 - 10 | 1122 | 117 | |
NM_001289752.1 | NP_001276681.1 | 9 - 10 | 1047 | 117 | |
GenBank mRNA | AB209703.1 | - | 10 - 11 | 1101 | 117 |
AK225301.1 | - | 10 - 11 | 1127 | 117 | |
AK294146.1 | - | 11 - 12 | 1128 | 117 | |
AK295080.1 | - | 8 - 9 | 985 | 117 | |
AK297508.1 | - | 9 - 10 | 1056 | 117 | |
AK300077.1 | - | 9 - 10 | 980 | 117 | |
AK312346.1 | - | 10 - 11 | 1138 | 117 | |
BC018648.2 | - | 10 - 11 | 1100 | 117 | |
BC018696.2 | - | 10 - 11 | 1100 | 117 | |
J03490.1 | - | 10 - 11 | 1123 | 117 | |
J03620.1 | - | 10 - 11 | 1097 | 117 |