The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000108.4 | NP_000099.2 | 6 - 7 | 586 | 140 |
NM_001289750.1 | NP_001276679.1 | 4 - 5 | 437 | 140 | |
NM_001289751.1 | NP_001276680.1 | 5 - 6 | 517 | 140 | |
GenBank mRNA | AB209703.1 | - | 6 - 7 | 496 | 140 |
AK225289.1 | - | 6 - 7 | 510 | 140 | |
AK225301.1 | - | 6 - 7 | 522 | 140 | |
AK294146.1 | - | 7 - 8 | 523 | 140 | |
AK295080.1 | - | 4 - 5 | 380 | 140 | |
AK310220.1 | - | 6 - 7 | 602 | 140 | |
AK312346.1 | - | 6 - 7 | 533 | 140 | |
BC018648.2 | - | 6 - 7 | 495 | 140 | |
BC018696.2 | - | 6 - 7 | 495 | 140 | |
J03490.1 | - | 6 - 7 | 518 | 140 | |
J03620.1 | - | 6 - 7 | 492 | 140 |