The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012081.5 | NP_036213.2 | 10 - 11 | 2111 | 146 |
XM_006714575.2 | XP_006714638.1 | 11 - 12 | 1951 | 146 | |
XM_017009240.1 | XP_016864729.1 | 11 - 12 | 2378 | 146 | |
XM_017009241.1 | XP_016864730.1 | 9 - 10 | 2206 | 146 | |
XM_017009242.1 | XP_016864731.1 | 10 - 11 | 1819 | 146 | |
XM_017009243.1 | XP_016864732.1 | 10 - 11 | 2422 | 146 | |
GenBank mRNA | AB209343.1 | - | 7 - 8 | 1400 | 146 |
AK296870.1 | - | 4 - 5 | 979 | 146 | |
AK297956.1 | - | 7 - 8 | 1140 | 146 | |
AK300310.1 | - | 4 - 5 | 896 | 146 | |
BC028412.2 | - | 10 - 11 | 2113 | 146 | |
BX538289.1 | - | 3 - 4 | 770 | 146 |