Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELL2
Entrez Gene ID
22936
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
-
Chromosome Location
Chr.5: 95885098 - 95962288 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012081.5 NP_036213.2 11 - 12 2154 134
XM_006714575.2 XP_006714638.1 12 - 13 1994 134
XM_017009240.1 XP_016864729.1 12 - 13 2421 134
XM_017009241.1 XP_016864730.1 10 - 11 2249 134
XM_017009242.1 XP_016864731.1 11 - 12 1862 134
XM_017009243.1 XP_016864732.1 11 - 12 2465 134
GenBank mRNA AB209343.1 - 8 - 9 1443 134
AK296870.1 - 5 - 6 1022 134
AK297956.1 - 8 - 9 1183 134
AK300310.1 - 5 - 6 939 134
BC028412.2 - 11 - 12 2156 134

View Full Product Details