Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPZ1
Entrez Gene ID
22818
Gene Name
coatomer protein complex subunit zeta 1
Gene Aliases
CGI-120, COPZ, HSPC181, zeta-COP, zeta1-COP
Chromosome Location
Chr.12: 54325090 - 54351851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271735.1 NP_001258664.1 1 - 2 94 82
NM_016057.2 NP_057141.1 1 - 2 94 82
NR_073424.1 1 - 2 94 82
XM_017018997.1 XP_016874486.1 1 - 2 75 82
GenBank mRNA AF151878.1 - 1 - 2 57 82
AF161529.1 - 1 - 2 54 82
AK025956.1 - 1 - 2 53 82
AK026190.1 - 1 - 2 53 82
AK295325.1 - 1 - 2 45 82
CB127540.1 - 1 - 2 81
DA562746.1 - 1 - 2 45 82

View Full Product Details