Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD6
Entrez Gene ID
84181
Gene Name
chromodomain helicase DNA binding protein 6
Gene Aliases
CHD-6, CHD5, RIGB
Chromosome Location
Chr.20: 41402101 - 41618494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032221.4 NP_115597.3 7 - 8 1158 74
XM_005260573.2 XP_005260630.1 6 - 7 980 74
XM_011529080.2 XP_011527382.1 6 - 7 1219 74
XM_017028099.1 XP_016883588.1 6 - 7 2654 74
XM_017028100.1 XP_016883589.1 6 - 7 1179 74
XM_017028101.1 XP_016883590.1 6 - 7 2654 74
XM_017028102.1 XP_016883591.1 6 - 7 2653 74
XM_017028103.1 XP_016883592.1 4 - 5 613 74
XM_017028104.1 XP_016883593.1 6 - 7 2654 74
GenBank mRNA AF525085.1 - 2 - 3 88 74
AK296955.1 - 6 - 7 988 74
AY034072.1 - 7 - 8 1158 74

View Full Product Details