Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTPAL
Entrez Gene ID
79183
Gene Name
alpha tocopherol transfer protein like
Gene Aliases
C20orf121
Chromosome Location
Chr.20: 44475886 - 44494603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039199.2 NP_001034288.1 4 - 5 873 121
NM_001261839.1 NP_001248768.1 4 - 5 771 121
NM_024331.4 NP_077307.2 5 - 6 897 121
XM_011529043.2 XP_011527345.1 5 - 6 1009 121
XM_011529044.2 XP_011527346.1 5 - 6 1624 121
XM_017028063.1 XP_016883552.1 5 - 6 786 121
GenBank mRNA AK307236.1 - 2 - 3 589 121
AK314045.1 - 4 - 5 832 121
BC003071.2 - 5 - 6 876 121
CR936621.1 - 4 - 5 827 121

View Full Product Details