The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012197.3 | NP_036329.3 | 16 - 17 | 2240 | 103 |
XM_011518440.2 | XP_011516742.1 | 16 - 17 | 2244 | 103 | |
XM_011518441.2 | XP_011516743.1 | 17 - 18 | 2312 | 103 | |
XM_011518442.2 | XP_011516744.1 | 15 - 16 | 2067 | 103 | |
XM_017014567.1 | XP_016870056.1 | 17 - 18 | 2529 | 103 | |
XM_017014568.1 | XP_016870057.1 | 17 - 18 | 2280 | 103 | |
XM_017014569.1 | XP_016870058.1 | 16 - 17 | 2342 | 103 | |
XM_017014570.1 | XP_016870059.1 | 15 - 16 | 1973 | 103 | |
GenBank mRNA | AB449897.1 | - | 15 - 16 | 2106 | 103 |
AJ011679.2 | - | 16 - 17 | 2211 | 103 | |
AK131449.1 | - | 18 - 19 | 2738 | 103 | |
AK294531.1 | - | 4 - 5 | 546 | 103 | |
BC054492.1 | - | 16 - 17 | 2220 | 103 |