The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144717.3 | NP_653318.2 | 2 - 3 | 464 | 74 |
XM_006713665.3 | XP_006713728.1 | 2 - 3 | 1193 | 74 | |
XM_011512910.2 | XP_011511212.1 | 3 - 4 | 368 | 74 | |
XM_011512911.2 | XP_011511213.1 | 2 - 3 | 321 | 74 | |
XM_017006632.1 | XP_016862121.1 | 1 - 2 | 277 | 74 | |
GenBank mRNA | AK296832.1 | - | 2 - 3 | 336 | 74 |
AK316084.1 | - | 2 - 3 | 344 | 74 | |
AY358305.1 | - | 2 - 3 | 464 | 74 | |
BC027449.1 | - | 1 - 2 | 119 | 74 | |
BC033292.1 | - | 2 - 3 | 455 | 74 | |
BC063696.1 | - | 2 - 3 | 328 | 74 |