Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMNAT3
Entrez Gene ID
349565
Gene Name
nicotinamide nucleotide adenylyltransferase 3
Gene Aliases
FKSG76, PNAT-3, PNAT3
Chromosome Location
Chr.3: 139560180 - 139678050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006341.1 XP_016861830.1 6 - 7 899 64
XM_017006342.1 XP_016861831.1 7 - 8 1005 64
XM_017006343.1 XP_016861832.1 6 - 7 965 64
XM_017006344.1 XP_016861833.1 5 - 6 865 64

View Full Product Details