The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099.4 | NP_001090.2 | 7 - 8 | 871 | 143 |
NM_001134194.1 | NP_001127666.1 | 7 - 8 | 871 | 143 | |
NM_001292037.1 | NP_001278966.1 | 6 - 7 | 772 | 143 | |
XM_011512946.1 | XP_011511248.1 | 7 - 8 | 835 | 143 | |
XM_011512947.2 | XP_011511249.1 | 6 - 7 | 736 | 143 | |
GenBank mRNA | AB102888.1 | - | 6 - 7 | 682 | 143 |
AK300540.1 | - | 6 - 7 | 733 | 143 | |
BC007460.1 | - | 7 - 8 | 831 | 143 | |
BC008493.1 | - | 7 - 8 | 828 | 143 | |
BC016344.1 | - | 7 - 8 | 835 | 143 | |
M24902.1 | - | 7 - 8 | 804 | 143 | |
M34840.1 | - | 7 - 8 | 795 | 143 | |
X52174.1 | - | 7 - 8 | 804 | 143 | |
X53605.1 | - | 7 - 8 | 817 | 143 |