The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000446.5 | NP_000437.3 | 6 - 7 | 790 | 95 |
GenBank mRNA | AK314027.1 | - | 6 - 7 | 716 | 95 |
BC074719.2 | - | 6 - 7 | 722 | 95 | |
D84371.1 | - | 6 - 7 | 747 | 95 | |
M63012.1 | - | 6 - 7 | 702 | 95 | |
M63013.1 | - | 6 - 7 | 641 | 95 | |
M63014.1 | - | 5 - 6 | 607 | 95 | |
S64615.1 | - | 6 - 7 | 702 | 95 | |
U53784.1 | - | 6 - 7 | 747 | 95 | |
Z70723.1 | - | 6 - 7 | 747 | 95 |