The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256267.1 | NP_001243196.1 | 2 - 3 | 189 | 115 |
NM_032578.3 | NP_115967.2 | 1 - 2 | 294 | 115 | |
NR_045663.3 | 1 - 2 | 294 | 115 | ||
GenBank mRNA | AF328296.1 | - | 1 - 2 | 489 | 115 |
AK027343.1 | - | 1 - 2 | 232 | 115 | |
AL832002.2 | - | 1 - 2 | 217 | 115 | |
AL834247.2 | - | 1 - 2 | 216 | 114 | |
BC142609.1 | - | 1 - 2 | 166 | 115 | |
BC144334.1 | - | 1 - 2 | 56 | 115 | |
BX647637.1 | - | 1 - 2 | 226 | 115 | |
BX647640.1 | - | 2 - 3 | 183 | 115 | |
BX648564.1 | - | 3 - 4 | 622 | 115 |