The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002922.3 | NP_002913.3 | 1 - 2 | 205 | 145 |
GenBank mRNA | AF493925.1 | - | 1 - 2 | 100 | 145 |
AK093544.1 | - | 1 - 2 | 157 | 145 | |
AK303138.1 | - | 1 - 2 | 159 | 145 | |
AK315607.1 | - | 1 - 2 | 159 | 145 | |
AL039059.1 | - | 1 - 2 | 209 | 145 | |
BC015510.1 | - | 1 - 2 | 137 | 145 | |
BG536972.1 | - | 1 - 2 | 205 | 145 | |
BT006668.1 | - | 1 - 2 | 100 | 145 | |
EU668340.1 | - | 1 - 2 | 137 | 145 | |
X73427.1 | - | 1 - 2 | 114 | 145 |