The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290321.2 | NP_001277250.1 | 19 - 20 | 5184 | 64 |
NM_001290322.2 | NP_001277251.1 | 19 - 20 | 6492 | 64 | |
NM_005509.5 | NP_005500.4 | 19 - 20 | 5184 | 64 | |
XM_005271909.4 | XP_005271966.1 | 20 - 21 | 4882 | 64 | |
XM_005271910.4 | XP_005271967.1 | 19 - 20 | 5227 | 64 | |
XM_005271912.2 | XP_005271969.1 | 20 - 21 | 4866 | 64 | |
XM_011543212.2 | XP_011541514.1 | 19 - 20 | 5227 | 64 | |
XM_011543213.2 | XP_011541515.1 | 20 - 21 | 4812 | 64 | |
XM_011543214.2 | XP_011541516.1 | 16 - 17 | 4393 | 64 | |
XM_011543215.2 | XP_011541517.1 | 15 - 16 | 4229 | 64 | |
XM_017009141.1 | XP_016864630.1 | 20 - 21 | 4882 | 64 | |
XM_017009142.1 | XP_016864631.1 | 20 - 21 | 4812 | 64 | |
XM_017009143.1 | XP_016864632.1 | 21 - 22 | 5061 | 64 | |
XM_017009144.1 | XP_016864633.1 | 19 - 20 | 4764 | 64 | |
XM_017009145.1 | XP_016864634.1 | 19 - 20 | 4634 | 64 | |
XM_017009146.1 | XP_016864635.1 | 19 - 20 | 6076 | 64 | |
XM_017009147.1 | XP_016864636.1 | 19 - 20 | 5227 | 64 | |
XM_017009148.1 | XP_016864637.1 | 18 - 19 | 5030 | 64 | |
GenBank mRNA | AB593142.1 | - | 19 - 20 | 6495 | 64 |
AJ005821.1 | - | 19 - 20 | 4781 | 64 |