Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
Chromosome Location
Chr.5: 119071002 - 119250170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290321.2 NP_001277250.1 32 - 33 8225 66
NM_001290322.2 NP_001277251.1 32 - 33 9533 66
NM_005509.5 NP_005500.4 32 - 33 8225 66
XM_005271909.4 XP_005271966.1 34 - 35 8037 66
XM_005271910.4 XP_005271967.1 33 - 34 8382 66
XM_005271912.2 XP_005271969.1 34 - 35 8021 66
XM_011543212.2 XP_011541514.1 33 - 34 8382 66
XM_011543213.2 XP_011541515.1 34 - 35 7967 66
XM_011543214.2 XP_011541516.1 30 - 31 7548 66
XM_011543215.2 XP_011541517.1 29 - 30 7384 66
XM_017009141.1 XP_016864630.1 33 - 34 7923 66
XM_017009142.1 XP_016864631.1 33 - 34 7853 66
XM_017009143.1 XP_016864632.1 35 - 36 8216 66
XM_017009144.1 XP_016864633.1 33 - 34 7919 66
XM_017009145.1 XP_016864634.1 33 - 34 7789 66
XM_017009146.1 XP_016864635.1 33 - 34 9231 66
XM_017009147.1 XP_016864636.1 33 - 34 8382 66
XM_017009148.1 XP_016864637.1 32 - 33 8185 66
GenBank mRNA AB593142.1 - 32 - 33 9536 66
AJ005821.1 - 32 - 33 7822 66
BC144537.1 - 32 - 33 7768 66
BC150616.1 - 32 - 33 7768 66

View Full Product Details