Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
Chromosome Location
Chr.5: 119071002 - 119250170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290321.2 NP_001277250.1 38 - 39 8811 130
NM_001290322.2 NP_001277251.1 37 - 38 10056 130
NM_005509.5 NP_005500.4 37 - 38 8748 130
XM_005271909.4 XP_005271966.1 40 - 41 8623 130
XM_005271910.4 XP_005271967.1 38 - 39 8905 130
XM_005271912.2 XP_005271969.1 40 - 41 8607 130
XM_011543212.2 XP_011541514.1 39 - 40 8968 130
XM_011543213.2 XP_011541515.1 40 - 41 8553 130
XM_011543214.2 XP_011541516.1 36 - 37 8134 130
XM_011543215.2 XP_011541517.1 35 - 36 7970 130
XM_017009141.1 XP_016864630.1 39 - 40 8509 130
XM_017009142.1 XP_016864631.1 38 - 39 8376 130
XM_017009143.1 XP_016864632.1 41 - 42 8802 130
XM_017009144.1 XP_016864633.1 39 - 40 8505 130
XM_017009145.1 XP_016864634.1 39 - 40 8375 130
XM_017009146.1 XP_016864635.1 39 - 40 9817 130
XM_017009147.1 XP_016864636.1 39 - 40 8968 130
XM_017009148.1 XP_016864637.1 38 - 39 8771 130
GenBank mRNA AB593142.1 - 37 - 38 10059 130
AJ005821.1 - 37 - 38 8345 130
BC144537.1 - 38 - 39 8354 130
BC150616.1 - 38 - 39 8354 130
BX649064.1 - 6 - 7 802 130

View Full Product Details