Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD10
Entrez Gene ID
51397
Gene Name
COMM domain containing 10
Gene Aliases
PTD002
Chromosome Location
Chr.5: 116084592 - 116446462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308080.1 NP_001295009.1 6 - 7 666 88
NM_016144.3 NP_057228.1 6 - 7 639 88
XM_017009553.1 XP_016865042.1 6 - 7 761 88
GenBank mRNA AF078857.1 - 6 - 7 594 88
AF161423.1 - 6 - 7 590 88
AK002147.1 - 6 - 7 615 88
AY542165.1 - 6 - 7 562 88
BC005179.1 - 6 - 7 585 88
BI859871.1 - 6 - 7 88

View Full Product Details