The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000347.5 | NP_000338.3 | 23 - 24 | 5027 | 68 |
NM_001024858.2 | NP_001020029.1 | 23 - 24 | 5027 | 68 | |
XM_005268023.4 | XP_005268080.1 | 24 - 25 | 5094 | 68 | |
XM_017021612.1 | XP_016877101.1 | 25 - 26 | 5242 | 68 | |
XM_017021613.1 | XP_016877102.1 | 24 - 25 | 5094 | 68 | |
XM_017021614.1 | XP_016877103.1 | 24 - 25 | 5094 | 68 | |
GenBank mRNA | AB209415.1 | - | 24 - 25 | 5116 | 68 |
AF465439.1 | - | 5 - 6 | 968 | 68 | |
BC136284.1 | - | 23 - 24 | 5006 | 68 | |
BC136285.1 | - | 23 - 24 | 5006 | 68 | |
J05500.1 | - | 24 - 25 | 5068 | 68 | |
M37884.1 | - | 9 - 10 | 1811 | 68 | |
M57948.1 | - | 10 - 11 | 2194 | 68 |