Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
Chromosome Location
Chr.14: 99511266 - 99604390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144995.1 NP_001138467.1 2 - 3 1298 59

View Full Product Details