The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017696.2 | NP_060166.2 | 2 - 3 | 629 | 131 |
NM_153255.4 | NP_694987.1 | 3 - 4 | 901 | 131 | |
GenBank mRNA | AK122665.1 | - | 3 - 4 | 741 | 131 |
BC031658.1 | - | 3 - 4 | 853 | 131 | |
KU178895.1 | - | 2 - 3 | 614 | 131 | |
KU178896.1 | - | 3 - 4 | 705 | 131 |