Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP24
Entrez Gene ID
23358
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
-
Chromosome Location
Chr.1: 55066359 - 55215374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015306.2 NP_056121.2 59 - 60 7334 81
XM_005270690.3 XP_005270747.1 46 - 47 5375 81
XM_017000831.1 XP_016856320.1 60 - 61 7399 81
XM_017000832.1 XP_016856321.1 60 - 61 7396 81
XM_017000833.1 XP_016856322.1 60 - 61 6891 81
XM_017000834.1 XP_016856323.1 59 - 60 6831 81
GenBank mRNA AB028980.1 - 18 - 19 2153 81
AK000321.1 - 3 - 4 318 81
AL832370.1 - 21 - 22 2560 81
BC029660.1 - 4 - 5 387 81

View Full Product Details