Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDTC1
Entrez Gene ID
23038
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
ADP, DCAF9
Chromosome Location
Chr.1: 27234516 - 27308633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276252.1 NP_001263181.1 14 - 15 2183 78
NM_015023.4 NP_055838.2 14 - 15 2180 78
XM_011541057.1 XP_011539359.1 14 - 15 1861 78
GenBank mRNA AK023101.1 - 5 - 6 722 78
AK023778.1 - 5 - 6 762 78
AK315564.1 - 14 - 15 2067 78
BC032523.1 - 14 - 15 2043 78

View Full Product Details