The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000795.3 | NP_000786.1 | 6 - 7 | 1044 | 96 |
XM_017017296.1 | XP_016872785.1 | 6 - 7 | 1026 | 96 | |
GenBank mRNA | AB209832.1 | - | 5 - 6 | 839 | 96 |
BC021195.2 | - | 6 - 7 | 904 | 96 | |
M29066.1 | - | 5 - 6 | 842 | 96 | |
M30625.1 | - | 6 - 7 | 1145 | 96 | |
X51362.1 | - | 6 - 7 | 974 | 96 | |
X51645.1 | - | 5 - 6 | 837 | 96 |