The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130991.2 | NP_001124463.1 | 12 - 13 | 1442 | 91 |
NM_006389.4 | NP_006380.1 | 12 - 13 | 1575 | 91 | |
XM_005271392.3 | XP_005271449.1 | 12 - 13 | 1436 | 91 | |
XM_005271393.3 | XP_005271450.1 | 12 - 13 | 1416 | 91 | |
XM_005271394.3 | XP_005271451.1 | 12 - 13 | 1483 | 91 | |
XM_017017095.1 | XP_016872584.1 | 12 - 13 | 1471 | 91 | |
XM_017017096.1 | XP_016872585.1 | 12 - 13 | 1492 | 91 | |
XM_017017097.1 | XP_016872586.1 | 12 - 13 | 1415 | 91 | |
GenBank mRNA | AK127324.1 | - | 11 - 12 | 1444 | 91 |
AK294905.1 | - | 13 - 14 | 1603 | 91 | |
AK301514.1 | - | 10 - 11 | 1134 | 91 | |
AK304264.1 | - | 11 - 12 | 1374 | 91 | |
AK307624.1 | - | 10 - 11 | 1057 | 91 | |
AK314178.1 | - | 12 - 13 | 1466 | 91 | |
BC017726.1 | - | 12 - 13 | 1393 | 91 | |
BC072436.1 | - | 12 - 13 | 1441 | 91 | |
DQ350134.1 | - | 12 - 13 | 1463 | 91 | |
DQ372932.1 | - | 12 - 13 | 1463 | 91 | |
U65785.1 | - | 12 - 13 | 1443 | 91 |