Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
Chromosome Location
Chr.11: 119044185 - 119057246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130991.2 NP_001124463.1 23 - 24 2939 91
NM_006389.4 NP_006380.1 23 - 24 3072 91
XM_005271392.3 XP_005271449.1 23 - 24 2933 91
XM_005271393.3 XP_005271450.1 23 - 24 2913 91
XM_005271394.3 XP_005271451.1 23 - 24 2980 91
XM_017017095.1 XP_016872584.1 23 - 24 2968 91
XM_017017096.1 XP_016872585.1 23 - 24 2989 91
XM_017017097.1 XP_016872586.1 23 - 24 2912 91
GenBank mRNA AK127324.1 - 22 - 23 2941 91
AK222756.1 - 23 - 24 2965 91
AK294905.1 - 23 - 24 2914 91
AK299648.1 - 16 - 17 1863 91
AK301514.1 - 21 - 22 2631 91
AK314178.1 - 23 - 24 2963 91
DQ350134.1 - 23 - 24 2960 91
DQ372932.1 - 23 - 24 2960 91
U65785.1 - 23 - 24 2940 91

View Full Product Details