The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005181.2 | NP_001005181.1 | 1 - 1 | 136 | 74 |
GenBank mRNA | BC136879.1 | - | 1 - 1 | 136 | 74 |
BC136880.1 | - | 1 - 1 | 136 | 74 |