Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
Chromosome Location
Chr.3: 58716415 - 59050078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198463.2 NP_940865.1 4 - 5 309 98
XM_005264929.2 XP_005264986.1 4 - 5 689 98
XM_006713013.3 XP_006713076.1 4 - 5 689 98
XM_011533451.2 XP_011531753.1 4 - 5 689 98
XM_011533453.2 XP_011531755.1 4 - 5 689 98
XM_011533455.2 XP_011531757.1 4 - 5 689 98
XM_011533456.1 XP_011531758.1 4 - 5 398 98
XM_011533459.2 XP_011531761.1 4 - 5 689 98
XM_017005849.1 XP_016861338.1 4 - 5 689 98
XM_017005850.1 XP_016861339.1 4 - 5 367 98
XM_017005851.1 XP_016861340.1 4 - 5 692 98
GenBank mRNA AK124111.1 - 4 - 5 311 98
AK296111.1 - 4 - 5 403 98
BC050317.1 - 4 - 5 676 98
BC132815.1 - 2 - 3 123 98
BC151142.1 - 2 - 3 123 98

View Full Product Details