Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEG1
Entrez Gene ID
57493
Gene Name
heart development protein with EGF like domains 1
Gene Aliases
HEG, MST112, MSTP112
Chromosome Location
Chr.3: 124965710 - 125055958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020733.1 NP_065784.1 13 - 14 3763 74
XM_005247666.1 XP_005247723.1 14 - 15 4058 74
GenBank mRNA AB033063.2 - 12 - 13 3358 74
AK074987.1 - 3 - 4 349 74
BC004539.2 - 3 - 4 279 74

View Full Product Details