Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
Chromosome Location
Chr.11: 113386014 - 113400418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178510.1 NP_848605.1 1 - 2 284 85
XM_011542736.2 XP_011541038.1 1 - 2 2061 85
XM_011542737.2 XP_011541039.1 1 - 2 2061 85
XM_017017475.1 XP_016872964.1 1 - 2 2061 85
GenBank mRNA AF487542.1 - 1 - 2 190 85
AJ541797.2 - 1 - 2 284 85

View Full Product Details