Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
Chromosome Location
Chr.11: 113386014 - 113400418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178510.1 NP_848605.1 3 - 4 732 80
XM_011542737.2 XP_011541039.1 3 - 4 2509 80
GenBank mRNA AF487542.1 - 3 - 4 638 80
AJ541797.2 - 3 - 4 732 80

View Full Product Details