Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C10orf67
Entrez Gene ID
256815
Gene Name
chromosome 10 open reading frame 67
Gene Aliases
C10orf115, LINC01552, bA215C7.4
Chromosome Location
Chr.10: 23202851 - 23345216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011519446.1 XP_011517748.1 4 - 5 992 82

View Full Product Details