The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178127.4 | NP_835228.2 | 1 - 2 | 502 | 163 |
XM_011542735.2 | XP_011541037.1 | 1 - 2 | 496 | 163 | |
XM_017017466.1 | XP_016872955.1 | 1 - 2 | 496 | 163 | |
GenBank mRNA | BC049170.1 | - | 1 - 2 | 505 | 163 |
BC053957.1 | - | 1 - 2 | 505 | 163 |