The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178127.4 | NP_835228.2 | 2 - 3 | 693 | 129 |
XM_011542735.2 | XP_011541037.1 | 2 - 3 | 687 | 129 | |
XM_017017466.1 | XP_016872955.1 | 2 - 3 | 687 | 129 | |
GenBank mRNA | AK291523.1 | - | 1 - 2 | 157 | 129 |
AY169281.1 | - | 1 - 2 | 151 | 129 | |
AY358827.1 | - | 1 - 2 | 141 | 129 | |
BC049170.1 | - | 2 - 3 | 696 | 129 | |
BC053957.1 | - | 2 - 3 | 696 | 129 |