Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCC1
Entrez Gene ID
6599
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
Gene Aliases
BAF155, CRACC1, Rsc8, SRG3, SWI3
Chromosome Location
Chr.3: 47585888 - 47781915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003074.3 NP_003065.3 10 - 11 1153 214
XM_011534034.1 XP_011532336.1 10 - 11 884 214
GenBank mRNA AK302541.1 - 10 - 11 928 214
BC021862.1 - 10 - 11 1090 214
BC032564.1 - 10 - 11 1137 214
BC039843.1 - 10 - 11 1057 214
BC050564.1 - 10 - 11 1082 214
BC065253.1 - 10 - 11 1122 214
BC113465.1 - 10 - 11 1101 214
BC117213.1 - 10 - 11 1101 214
U66615.1 - 10 - 11 1089 214

View Full Product Details