The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130991.2 | NP_001124463.1 | 1 - 2 | 93 | 60 |
XM_005271392.3 | XP_005271449.1 | 1 - 2 | 87 | 60 | |
GenBank mRNA | AK299648.1 | - | 1 - 2 | 49 | 60 |
BC004560.2 | - | 1 - 2 | 41 | 60 | |
BP236985.1 | - | 1 - 2 | 61 | 60 | |
HY089857.1 | - | 1 - 2 | 94 | 60 |