Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMAT1
Entrez Gene ID
84460
Gene Name
zinc finger matrin-type 1
Gene Aliases
-
Chromosome Location
Chr.X: 101882288 - 101932135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282400.1 NP_001269329.1 8 - 9 1408 149
NM_001282401.1 NP_001269330.1 7 - 8 1355 149
GenBank mRNA AK094141.1 - 8 - 9 1369 149
AK296520.1 - 7 - 8 1320 149

View Full Product Details