The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145158.1 | NP_001138630.1 | 9 - 10 | 1383 | 73 |
NM_006620.3 | NP_006611.1 | 10 - 11 | 1509 | 73 | |
GenBank mRNA | AB028961.1 | - | 5 - 6 | 738 | 73 |
AJ459826.1 | - | 10 - 11 | 1495 | 73 | |
AK024258.1 | - | 10 - 11 | 1430 | 73 | |
AK293573.1 | - | 7 - 8 | 717 | 73 | |
AK295545.1 | - | 9 - 10 | 1304 | 73 | |
AK298336.1 | - | 7 - 8 | 912 | 73 | |
BC001465.2 | - | 10 - 11 | 1418 | 73 | |
BC040849.1 | - | 10 - 11 | 1413 | 73 | |
BX648033.1 | - | 7 - 8 | 1174 | 73 | |
U87791.1 | - | 10 - 11 | 1496 | 73 |