Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
Chromosome Location
Chr.15: 76059929 - 76204963 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152335.2 NP_689548.2 6 - 7 674 71
XM_005254160.2 XP_005254217.1 6 - 7 597 71
XM_017021915.1 XP_016877404.1 8 - 9 890 71
XM_017021916.1 XP_016877405.1 8 - 9 836 71
XM_017021917.1 XP_016877406.1 8 - 9 822 71
GenBank mRNA AK095509.1 - 6 - 7 566 71
AK308676.1 - 5 - 6 492 71
BX398159.2 - 5 - 6 590 71
DA483185.1 - 6 - 7 566 71
DR007145.1 - 2 - 3 144 71

View Full Product Details