The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199260.1 | NP_001186189.1 | 5 - 6 | 1366 | 112 |
NM_145034.4 | NP_659471.1 | 4 - 5 | 947 | 112 | |
XM_005244939.4 | XP_005244996.1 | 5 - 6 | 1339 | 112 | |
XM_017000474.1 | XP_016855963.1 | 6 - 7 | 1484 | 112 | |
XM_017000475.1 | XP_016855964.1 | 4 - 5 | 1208 | 112 | |
GenBank mRNA | AF464140.1 | - | 4 - 5 | 941 | 112 |
BC101532.1 | - | 2 - 3 | 651 | 112 | |
BC112225.1 | - | 2 - 3 | 651 | 112 |