Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PACS2
Entrez Gene ID
23241
Gene Name
phosphofurin acidic cluster sorting protein 2
Gene Aliases
PACS-2, PACS1L
Chromosome Location
Chr.14: 105300811 - 105398147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100913.2 NP_001094383.2 22 - 23 2762 122
NM_001243127.2 NP_001230056.1 21 - 22 2239 122
NM_015197.3 NP_056012.2 21 - 22 2717 122
XM_006720090.1 XP_006720153.1 22 - 23 2430 122
XM_006720091.1 XP_006720154.1 21 - 22 2397 122
XM_006720092.2 XP_006720155.1 21 - 22 2358 122
XM_017021105.1 XP_016876594.1 22 - 23 2418 122
XM_017021106.1 XP_016876595.1 22 - 23 2415 122
XM_017021107.1 XP_016876596.1 22 - 23 2406 122
XM_017021108.1 XP_016876597.1 22 - 23 2403 122
XM_017021109.1 XP_016876598.1 21 - 22 2394 122
XM_017021110.1 XP_016876599.1 21 - 22 2385 122
XM_017021111.1 XP_016876600.1 21 - 22 2263 122
XM_017021112.1 XP_016876601.1 22 - 23 2198 122
GenBank mRNA AB011174.1 - 22 - 23 2433 122
AY320284.2 - 21 - 22 2716 122
BC010663.1 - 18 - 19 1824 122
BC050351.1 - 21 - 22 2255 122
BC065220.1 - 21 - 22 2330 122
BC131591.1 - 21 - 22 2217 122

View Full Product Details