The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256267.1 | NP_001243196.1 | 16 - 17 | 3345 | 76 |
NM_001256268.1 | NP_001243197.1 | 19 - 20 | 3754 | 76 | |
NM_032578.3 | NP_115967.2 | 15 - 16 | 3450 | 76 | |
NR_045662.3 | 15 - 16 | 2585 | 76 | ||
NR_045663.3 | 13 - 14 | 3287 | 76 | ||
XM_017016833.1 | XP_016872322.1 | 16 - 17 | 3786 | 76 | |
XM_017016834.1 | XP_016872323.1 | 15 - 16 | 3244 | 76 | |
GenBank mRNA | AF328296.1 | - | 15 - 16 | 3645 | 76 |
AL832002.2 | - | 15 - 16 | 3373 | 76 | |
AL832379.1 | - | 15 - 16 | 2547 | 76 | |
AL834247.2 | - | 15 - 16 | 3372 | 76 | |
BC144334.1 | - | 13 - 14 | 3049 | 76 | |
BX647637.1 | - | 15 - 16 | 3381 | 76 | |
BX647640.1 | - | 16 - 17 | 3338 | 76 |