The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002208.4 | NP_002199.3 | 27 - 28 | 3233 | 69 |
XM_011523823.2 | XP_011522125.1 | 27 - 28 | 3734 | 69 | |
XM_011523827.2 | XP_011522129.1 | 26 - 27 | 3564 | 69 | |
XM_011523828.2 | XP_011522130.1 | 25 - 26 | 3490 | 69 | |
XM_017024586.1 | XP_016880075.1 | 27 - 28 | 5041 | 69 | |
GenBank mRNA | BC113436.1 | - | 27 - 28 | 3223 | 69 |
BC117207.1 | - | 27 - 28 | 3223 | 69 | |
L25851.2 | - | 27 - 28 | 3259 | 69 |