The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006366.2 | NP_006357.1 | 5 - 6 | 689 | 69 |
XM_011514233.2 | XP_011512535.1 | 4 - 5 | 900 | 69 | |
GenBank mRNA | AK297538.1 | - | 4 - 5 | 405 | 69 |
AK308461.1 | - | 4 - 5 | 404 | 69 | |
AK312362.1 | - | 5 - 6 | 817 | 69 | |
BC008481.1 | - | 5 - 6 | 482 | 69 | |
CR457022.1 | - | 4 - 5 | 446 | 69 | |
U02390.1 | - | 5 - 6 | 529 | 69 |