Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD29
Entrez Gene ID
147463
Gene Name
ankyrin repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.18: 23598926 - 23662914 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308238.1 NP_001295167.1 3 - 4 386 66
NM_173505.3 NP_775776.2 3 - 4 386 66
XM_011525831.2 XP_011524133.1 2 - 3 2326 66
XM_011525832.2 XP_011524134.1 2 - 3 2444 66
GenBank mRNA AK057782.1 - 3 - 4 316 66
AK313667.1 - 3 - 4 304 66
BC030622.1 - 3 - 4 391 66

View Full Product Details