Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNF4A
Entrez Gene ID
3172
Gene Name
hepatocyte nuclear factor 4 alpha
Gene Aliases
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
Chromosome Location
Chr.20: 44355801 - 44432845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030003.2 NP_001025174.1 1 - 2 48 82
NM_001030004.2 NP_001025175.1 1 - 2 48 82
NM_175914.4 NP_787110.2 1 - 2 48 82
GenBank mRNA AY680696.1 - 1 - 2 48 82
AY680697.1 - 1 - 2 48 82
AY680698.1 - 1 - 2 48 82

View Full Product Details