The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199377.1 | NP_001186306.1 | 6 - 7 | 941 | 60 |
NM_015162.4 | NP_055977.3 | 6 - 7 | 953 | 60 | |
XM_011521390.2 | XP_011519692.1 | 6 - 7 | 767 | 60 | |
XM_011521391.2 | XP_011519693.2 | 5 - 6 | 2626 | 60 | |
XM_017022025.1 | XP_016877514.1 | 6 - 7 | 787 | 60 | |
XM_017022026.1 | XP_016877515.1 | 9 - 10 | 1588 | 60 | |
GenBank mRNA | AF179481.1 | - | 6 - 7 | 794 | 60 |
AK054882.1 | - | 6 - 7 | 803 | 60 | |
AK294202.1 | - | 2 - 3 | 239 | 60 | |
AK295269.1 | - | 6 - 7 | 759 | 60 | |
AK299597.1 | - | 5 - 6 | 719 | 60 | |
AK314508.1 | - | 6 - 7 | 953 | 60 | |
BC009289.2 | - | 6 - 7 | 752 | 60 |