Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPP7
Entrez Gene ID
143098
Gene Name
membrane palmitoylated protein 7
Gene Aliases
-
Chromosome Location
Chr.10: 28050993 - 28303066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318170.1 NP_001305099.1 3 - 4 482 88
NM_173496.4 NP_775767.2 5 - 6 508 88
NR_134517.1 3 - 4 482 88
XM_005252369.3 XP_005252426.1 3 - 4 440 88
XM_011519337.2 XP_011517639.1 4 - 5 407 88
GenBank mRNA AK315046.1 - 3 - 4 427 88
AL832380.1 - 2 - 3 278 88
BC038105.2 - 5 - 6 500 88

View Full Product Details