The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318170.1 | NP_001305099.1 | 4 - 5 | 573 | 124 |
NM_173496.4 | NP_775767.2 | 6 - 7 | 599 | 124 | |
NR_134517.1 | 4 - 5 | 573 | 124 | ||
NR_134518.1 | 3 - 4 | 454 | 124 | ||
XM_005252369.3 | XP_005252426.1 | 4 - 5 | 531 | 124 | |
XM_011519337.2 | XP_011517639.1 | 5 - 6 | 498 | 124 | |
XM_011519338.2 | XP_011517640.1 | 3 - 4 | 454 | 124 | |
XM_017015741.1 | XP_016871230.1 | 3 - 4 | 1211 | 124 | |
XM_017015742.1 | XP_016871231.1 | 4 - 5 | 512 | 124 | |
XM_017015743.1 | XP_016871232.1 | 3 - 4 | 1215 | 124 | |
GenBank mRNA | AK057360.2 | - | 4 - 5 | 496 | 124 |
AK301592.1 | - | 3 - 4 | 454 | 124 | |
AK315046.1 | - | 4 - 5 | 518 | 124 | |
AL832380.1 | - | 3 - 4 | 369 | 124 | |
BC038105.2 | - | 6 - 7 | 591 | 124 |