Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPHA8
Entrez Gene ID
2046
Gene Name
EPH receptor A8
Gene Aliases
EEK, EK3, HEK3
Chromosome Location
Chr.1: 22563508 - 22603594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011540969.1 XP_011539271.1 12 - 13 2218 66
XM_011540970.1 XP_011539272.1 10 - 11 1952 66
XM_011540972.1 XP_011539274.1 11 - 12 2119 66
XM_011540973.1 XP_011539275.1 9 - 10 1854 66
XM_011540975.2 XP_011539277.1 7 - 8 1030 66
GenBank mRNA AB040892.1 - 8 - 9 1476 66

View Full Product Details